Gene Transcripts

Gene Symbol
Zcchc16
Entrez Gene ID
619287
Gene Name
zinc finger, CCHC domain containing 16
Gene Aliases
C230031A03Rik, Mar4
Chromosome Location
Chr. X: 144688218 - 145122414 on Build GRCm38
UniGene ID
Mm.387961
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033795.4 NP_001028967.2 7 1070
GenBank mRNA AK136928.1 - 1 759
AK141066.1 - 7 1072
BC138958.1 - 1 346 8861980
BC138959.1 - 1 346 8861981

View Full Product Details