Gene Transcripts

Gene Symbol
Nedd4
Entrez Gene ID
25489
Gene Name
neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase
Gene Aliases
Nedd4a
Chromosome Location
Chr. 8: 79323408 - 79408842 on Build Rnor_6.0
UniGene ID
Rn.99540
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012986.1 NP_037118.1 13 1228
XM_008766214.1 XP_008764436.1 8 4639
GenBank mRNA CB615796.1 - 3 338
U50842.1 - 13 1154

View Full Product Details