Gene Transcripts

Gene Symbol
SPP1
Entrez Gene ID
6696
Gene Name
secreted phosphoprotein 1
Gene Aliases
BNSP, BSPI, ETA-1, OPN
Chromosome Location
Chr. 4: 87975650 - 87983411 on Build GRCh38
UniGene ID
Hs.313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000582.2 NP_000573.1 3 236
NM_001040058.1 NP_001035147.1 3 236
NM_001040060.1 NP_001035149.1 3 236
NM_001251829.1 NP_001238758.1 3 236
NM_001251830.1 NP_001238759.1 3 236
GenBank mRNA AB209987.1 - 2 71
AB469789.1 - 2 71
AF052124.1 - 3 158
AK057738.1 - 3 172
AK075463.1 - 3 172
AK290090.1 - 3 172
AK290104.1 - 3 173
AK295491.1 - 3 171
AK296035.1 - 3 172
AK315461.1 - 3 166
AY956318.1 - 3 138
BC007016.1 - 3 172 4052438
BC017387.1 - 3 176 3828885
BC022844.1 - 3 173 4284921
BC093033.1 - 3 228 5288027
BF699765.1 - 3 237
BX648003.1 - 3 190
CB117856.1 - 3 236
D28759.1 - 2 71
D28760.1 - 2 71
D28761.1 - 2 71
DA419665.1 - 3 236
DQ839491.1 - 2 71
DQ846870.1 - 2 71
DQ846871.1 - 2 71
J04765.1 - 3 179
JF412666.1 - 2 73
JF412667.1 - 3 234
KR062183.1 - 3 171
M83248.1 - 3 168
X13694.1 - 3 138

View Full Product Details