Gene Transcripts

Gene Symbol
SPP1
Entrez Gene ID
6696
Gene Name
secreted phosphoprotein 1
Gene Aliases
BNSP, BSPI, ETA-1, OPN
Chromosome Location
Chr. 4: 87975650 - 87983411 on Build GRCh38
UniGene ID
Hs.313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000582.2 NP_000573.1 5, 6 668
NM_001040058.1 NP_001035147.1 6, 7 710
NM_001040060.1 NP_001035149.1 5, 6 629
NM_001251829.1 NP_001238758.1 4, 5 587
NM_001251830.1 NP_001238759.1 7, 8 899
XM_017008564.1 XP_016864053.1 4, 5 1136
GenBank mRNA AB209987.1 - 3, 4 422
AB469789.1 - 3, 4 422
AF052124.1 - 5, 6 590
AK057738.1 - 6, 7 646
AK075463.1 - 7, 8 580
AK290090.1 - 5, 6 565
AK290104.1 - 6, 7 647
AK295491.1 - 7, 8 834
AK296035.1 - 5, 6 565
AK315461.1 - 5, 6 598
BC007016.1 - 5, 6 604 4052438
BC017387.1 - 6, 7 650 3828885
BC022844.1 - 5, 6 605 4284921
BC093033.1 - 5, 6 621 5288027
BX648003.1 - 5, 6 583
D28759.1 - 5, 6 545
D28760.1 - 4, 5 503
D28761.1 - 4, 5 464
DQ839491.1 - 5, 6 545
DQ846870.1 - 5, 6 545
DQ846871.1 - 5, 6 545
J04765.1 - 5, 6 611
JF412666.1 - 6, 7 481
JF412667.1 - 5, 6 627
KR062183.1 - 7, 8 834
M83248.1 - 6, 7 642
X13694.1 - 6, 7 612

View Full Product Details