Gene Transcripts

Gene Symbol
SSR2
Entrez Gene ID
6746
Gene Name
signal sequence receptor subunit 2
Gene Aliases
HSD25, TLAP, TRAP-BETA, TRAPB
Chromosome Location
Chr. 1: 156009048 - 156020992 on Build GRCh38
UniGene ID
Hs.74564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003145.3 NP_003136.1 4 405
XM_011509923.2 XP_011508225.1 4 430
GenBank mRNA AK222600.1 - 4 348
AK300678.1 - 4 405
AK313821.1 - 4 405
AY251536.1 - 4 365
BC000341.2 - 4 351 2822983
BP196192.1 - 4 405
BX649192.1 - 4 390
CR456973.1 - 3 325
D37991.1 - 4 349
X74104.1 - 4 375

View Full Product Details