Gene Transcripts

Gene Symbol
Dbx1
Entrez Gene ID
292934
Gene Name
developing brain homeobox 1
Gene Aliases
Chromosome Location
Chr. 1: 104969218 - 104973648 on Build Rnor_6.0
UniGene ID
Rn.104881
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009644.1 NP_001009644.1 1 129
GenBank mRNA AB197138.1 - 1 129

View Full Product Details