Gene Transcripts

Gene Symbol
Fam89b
Entrez Gene ID
309170
Gene Name
family with sequence similarity 89, member B
Gene Aliases
Mtvr2
Chromosome Location
Chr. 1: 221085894 - 221087449 on Build Rnor_6.0
UniGene ID
Rn.160645
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015013.1 NP_001015013.1 2 420
GenBank mRNA BC093377.1 - 2 420

View Full Product Details