Gene Transcripts

Gene Symbol
Coa3
Entrez Gene ID
498000
Gene Name
cytochrome C oxidase assembly factor 3
Gene Aliases
Ccdc56, RGD1564337
Chromosome Location
Chr. 10: 89198750 - 89199731 on Build Rnor_6.0
UniGene ID
Rn.32253
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109047.1 NP_001102517.1 2 445
GenBank mRNA FQ221438.1 - 2 450

View Full Product Details