Gene Transcripts

Gene Symbol
Spink13
Entrez Gene ID
689570
Gene Name
serine peptidase inhibitor, Kazal type 13 (putative)
Gene Aliases
Spink5l3
Chromosome Location
Chr. 18: 57935271 - 58081096 on Build Rnor_6.0
UniGene ID
Rn.218699
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109539.1 NP_001103009.1 4 311
XM_008772175.1 XP_008770397.1 4 233

View Full Product Details