Gene Transcripts

Gene Symbol
C16orf86
Entrez Gene ID
388284
Gene Name
chromosome 16 open reading frame 86
Gene Aliases
Chromosome Location
Chr. 16: 67666806 - 67670523 on Build GRCh38
UniGene ID
Hs.632208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012984.2 NP_001013002.2 3 607
XM_005255952.4 XP_005256009.1 3 615
GenBank mRNA AK123796.1 - 2 957
BC032064.1 - 3 584
BC064498.1 - 3 653
BC082249.1 - 3 653
BC132878.1 - 1 522
BC132880.1 - 1 522

View Full Product Details