Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027179.1 | NP_081455.1 | 9 | 720 | |
NM_134118.5 | NP_598879.1 | 10 | 765 | ||
GenBank mRNA | AK002384.1 | - | 10 | 735 | |
AK010513.1 | - | 9 | 656 | ||
AK010984.1 | - | 10 | 739 | ||
AK013558.1 | - | 10 | 718 | ||
AK020708.1 | - | 9 | 716 | ||
AK171718.1 | - | 5 | 472 | ||
AK172506.1 | - | 5 | 450 | ||
AK189199.1 | - | 6 | 296 | ||
BC019984.1 | - | 10 | 751 | 4240045 | |
BC094046.1 | - | 10 | 865 | 5064002 |