Gene Transcripts

Gene Symbol
Cyhr1
Entrez Gene ID
54151
Gene Name
cysteine and histidine rich 1
Gene Aliases
1110031M01Rik, AU042374, Chrp
Chromosome Location
Chr. 15: 76643395 - 76660208 on Build GRCm38
UniGene ID
Mm.247752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019396.3 NP_062269.1 5 978
GenBank mRNA AJ251516.1 - 5 831
BC039646.1 - 3 371
BE653118.1 - 3 404
CN527531.1 - 2 123

View Full Product Details