Gene Transcripts
- Gene Symbol
- Hist1h2aa
- Entrez Gene ID
- 24828
- Gene Name
- histone cluster 1, H2aa
- Gene Aliases
- Th2a
- Chromosome Location
- Chr. 17: 43422454 - 43422846 on Build Rnor_6.0
- UniGene ID
- Rn.138107
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021839.1 |
NP_068611.1
|
1 |
61 |
|
View Full Product Details