Gene Transcripts

Gene Symbol
FAM89B
Entrez Gene ID
23625
Gene Name
family with sequence similarity 89 member B
Gene Aliases
LRAP25, MTVR, MTVR1
Chromosome Location
Chr. 11: 65572349 - 65574198 on Build GRCh38
UniGene ID
Hs.731854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098784.1 NP_001092254.1 2 1394
NM_001098785.1 NP_001092255.1 2 1384
NM_152832.2 NP_690045.1 2 1345
GenBank mRNA AF052151.1 - 2 1281
BC023991.2 - 3 1792 2819240
BC032373.1 - 2 1356 5123262
BQ927525.1 - 1 585

View Full Product Details