Gene Transcripts

Gene Symbol
BEX1
Entrez Gene ID
55859
Gene Name
brain expressed X-linked 1
Gene Aliases
BEX2, HBEX2, HGR74-h
Chromosome Location
Chr. X: 103062653 - 103064240 on Build GRCh38
UniGene ID
Hs.334370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018476.3 NP_060946.3 3 691
GenBank mRNA AF183416.1 - 1 453
AF220189.1 - 3 617
AF237783.1 - 3 622
AY833561.1 - 3 613
BC018615.2 - 1 457
BM465948.1 - 3 632

View Full Product Details