Gene Transcripts

Gene Symbol
OR2S2
Entrez Gene ID
56656
Gene Name
olfactory receptor family 2 subfamily S member 2 (gene/pseudogene)
Gene Aliases
OR37A, OST715
Chromosome Location
Chr. 9: 35957108 - 35958154 on Build GRCh38
UniGene ID
Hs.553540
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019897.2 NP_063950.2 1 303
GenBank mRNA BC104869.1 - 1 303
BC113651.1 - 1 303 8322703

View Full Product Details