Gene Transcripts

Gene Symbol
EVA1C
Entrez Gene ID
59271
Gene Name
eva-1 homolog C (C. elegans)
Gene Aliases
B18, B19, C21orf63, C21orf64, FAM176C, PRED34, SUE21
Chromosome Location
Chr. 21: 32411692 - 32515400 on Build GRCh38
UniGene ID
Hs.208358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286556.1 NP_001273485.1 1, 2 575
NM_058187.4 NP_478067.2 1, 2 575
NR_135467.1 1, 2 631
XM_005261019.4 XP_005261076.1 1, 2 696
XM_006724038.3 XP_006724101.1 1, 2 696
XM_017028418.1 XP_016883907.1 1, 2 696
XM_017028419.1 XP_016883908.1 1, 2 696
GenBank mRNA AF358258.1 - 1, 2 158
AY358787.1 - 1, 2 568
BC038710.1 - 1, 2 310 4813873

View Full Product Details