Gene Transcripts

Gene Symbol
POPDC3
Entrez Gene ID
64208
Gene Name
popeye domain containing 3
Gene Aliases
POP3, bA355M14.1
Chromosome Location
Chr. 6: 105157900 - 105179983 on Build GRCh38
UniGene ID
Hs.458336
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022361.4 NP_071756.2 4 1137
NR_024539.1 4 678
XM_011536067.2 XP_011534369.1 4 1297
XM_017011194.1 XP_016866683.1 4 2580
XM_017011195.1 XP_016866684.1 4 2040
GenBank mRNA AF204171.1 - 3 882
AK314101.1 - 4 1137
BC022323.2 - 3 987 4293961

View Full Product Details