Gene Transcripts
- Gene Symbol
- PBX4
- Entrez Gene ID
- 80714
- Gene Name
- PBX homeobox 4
- Gene Aliases
- Chromosome Location
- Chr. 19: 19561707 - 19618916 on Build GRCh38
- UniGene ID
- Hs.466257
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025245.2 |
NP_079521.1
|
5, 6 |
1053 |
|
NR_038198.1 |
|
5, 6 |
952 |
|
XM_006722911.3 |
XP_006722974.1
|
4, 5 |
634 |
|
XM_011528320.2 |
XP_011526622.1
|
6, 7 |
1099 |
|
XM_011528322.2 |
XP_011526624.1
|
4, 5 |
688 |
|
XM_017027326.1 |
XP_016882815.1
|
6, 7 |
734 |
|
XM_017027327.1 |
XP_016882816.1
|
6, 7 |
715 |
|
XM_017027328.1 |
XP_016882817.1
|
6, 7 |
1099 |
|
XM_017027329.1 |
XP_016882818.1
|
3, 4 |
524 |
|
GenBank mRNA |
AJ300182.1 |
- |
4, 5 |
634 |
|
AK097427.1 |
- |
5, 6 |
790 |
|
AK301631.1 |
- |
5, 6 |
717 |
|
BC141859.1 |
- |
5, 6 |
1053 |
8860384 |
BC143588.1 |
- |
5, 6 |
728 |
|
BC143589.1 |
- |
5, 6 |
739 |
|
View Full Product Details