Gene Transcripts

Gene Symbol
LEAP2
Entrez Gene ID
116842
Gene Name
liver enriched antimicrobial peptide 2
Gene Aliases
LEAP-2
Chromosome Location
Chr. 5: 132873666 - 132874890 on Build GRCh38
UniGene ID
Hs.337588
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052971.2 NP_443203.1 2 201
GenBank mRNA AJ306405.1 - 2 201
AJ409065.1 - 2 1545
AY052782.1 - 2 201
BC070199.1 - 2 199 4711742
BX443829.2 - 2 189

View Full Product Details