Gene Transcripts

Gene Symbol
NKAIN2
Entrez Gene ID
154215
Gene Name
Na+/K+ transporting ATPase interacting 2
Gene Aliases
FAM77B, NKAIP2, TCBA, TCBA1
Chromosome Location
Chr. 6: 123803840 - 124825652 on Build GRCh38
UniGene ID
Hs.656604
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040214.2 NP_001035304.1 5 855
NM_001300737.1 NP_001287666.1 6 919
NM_001300738.1 NP_001287667.1 6 869
NM_001300740.1 NP_001287669.1 5 493
NM_153355.4 NP_699186.2 4 654
XM_011535501.2 XP_011533803.1 5 743
XM_011535502.2 XP_011533804.1 5 510
XM_011535503.2 XP_011533805.1 4 780
XM_017010318.1 XP_016865807.1 4 778
XM_017010319.1 XP_016865808.1 4 542
GenBank mRNA AK055326.1 - 5 488
AK308927.1 - 6 507
AK309385.1 - 6 468
AK309407.1 - 2 230
AK309802.1 - 6 507
AK310763.1 - 4 329
AK315886.1 - 5 494
BC035062.2 - 6 621 5264304
DB174077.1 - 3 329

View Full Product Details