Gene Transcripts

Gene Symbol
ZSCAN30
Entrez Gene ID
100101467
Gene Name
zinc finger and SCAN domain containing 30
Gene Aliases
ZNF-WYM, ZNF397OS, ZNF917
Chromosome Location
Chr. 18: 35251058 - 35290261 on Build GRCh38
UniGene ID
Hs.619840
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001112734.3 NP_001106205.1 4 1819
NM_001166012.2 NP_001159484.1 5 2023
NM_001288711.1 NP_001275640.1 2 1116
XM_005258183.4 XP_005258240.1 4 1788
XM_006722371.3 XP_006722434.1 5 1952
XM_011525789.2 XP_011524091.1 1 1017
XM_017025515.1 XP_016881004.1 5 1971
XM_017025516.1 XP_016881005.1 4 1804
XM_017025517.1 XP_016881006.1 4 1798
XM_017025518.1 XP_016881007.1 4 1870
XM_017025519.1 XP_016881008.1 4 1775
XM_017025520.1 XP_016881009.1 3 1669
XM_017025521.1 XP_016881010.1 3 1622
XM_017025522.1 XP_016881011.1 3 1601
GenBank mRNA AK001503.1 - 1 136
AK057386.1 - 1 572
AK303447.1 - 5 2007
AL832180.1 - 1 327
AY234408.1 - 4 1782

View Full Product Details