Gene Transcripts

Gene Symbol
HIST1H2AA
Entrez Gene ID
221613
Gene Name
histone cluster 1, H2aa
Gene Aliases
H2AA, H2AFR, TH2A, bA317E16.2
Chromosome Location
Chr. 6: 25726063 - 25726562 on Build GRCh38
UniGene ID
Hs.406739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170745.3 NP_734466.1 1 145
GenBank mRNA BC062211.1 - 1 149

View Full Product Details