Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000169.2 | NP_000160.1 | 7 | 1322 | |
GenBank mRNA | AK222627.1 | - | 7 | 1234 | |
AK291095.1 | - | 7 | 1241 | ||
BC002689.2 | - | 7 | 1218 | 3609235 | |
BT006864.1 | - | 7 | 1212 | ||
D00039.1 | - | 7 | 1136 | ||
X05790.1 | - | 7 | 1235 | ||
X16889.1 | - | 7 | 1214 |