Gene Transcripts
- Gene Symbol
- MYH14
- Entrez Gene ID
- 79784
- Gene Name
- myosin, heavy chain 14, non-muscle
- Gene Aliases
- DFNA4, DFNA4A, FP17425, MHC16, MYH17, NMHC II-C, NMHC-II-C, PNMHH, myosin
- Chromosome Location
- Chr. 19: 50188186 - 50310544 on Build GRCh38
- UniGene ID
- Hs.467142
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077186.1 |
NP_001070654.1
|
27, 28 |
3636 |
|
NM_001145809.1 |
NP_001139281.1
|
28, 29 |
3735 |
|
NM_024729.3 |
NP_079005.3
|
26, 27 |
3612 |
|
XM_006723386.3 |
XP_006723449.1
|
27, 28 |
3697 |
|
XM_011527320.2 |
XP_011525622.1
|
27, 28 |
3714 |
|
XM_011527321.2 |
XP_011525623.1
|
26, 27 |
3690 |
|
XM_011527323.2 |
XP_011525625.1
|
27, 28 |
3704 |
|
GenBank mRNA |
AB111886.1 |
- |
17, 18 |
2294 |
|
AB290169.1 |
- |
28, 29 |
3705 |
|
AY165122.1 |
- |
27, 28 |
3612 |
|
AY203926.1 |
- |
5, 6 |
677 |
|
BC129803.1 |
- |
15, 16 |
1709 |
|
View Full Product Details