Gene Transcripts

Gene Symbol
HRCT1
Entrez Gene ID
646962
Gene Name
histidine rich carboxyl terminus 1
Gene Aliases
LGLL338, PRO537, UNQ338
Chromosome Location
Chr. 9: 35906192 - 35907141 on Build GRCh38
UniGene ID
Hs.208081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039792.1 NP_001034881.1 1 337
GenBank mRNA AY358408.1 - 1 337

View Full Product Details