Gene Transcripts

Gene Symbol
Aebp1
Entrez Gene ID
11568
Gene Name
AE binding protein 1
Gene Aliases
ACLP
Chromosome Location
Chr. 11: 5861853 - 5872225 on Build GRCm38
UniGene ID
Mm.3317
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF053943.1 - 21 3538
AK004712.1 - 17 2547
BC082577.1 - 21 3550 30648530
X80478.1 - 12 2268

View Full Product Details