Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207105.3 | NP_996988.2 | 1, 2 | 188 | |
GenBank mRNA | AF119251.1 | - | 1, 2 | 103 | |
AF119252.1 | - | 1, 2 | 103 | ||
AK077436.1 | - | 1, 2 | 176 | ||
AK170370.1 | - | 1, 2 | 170 | ||
AY303784.1 | - | 1 | 21 | ||
AY303785.1 | - | 12 | |||
BY205244.1 | - | 1, 2 | 207 |