Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018766.2 | NP_061236.1 | 3, 4 | 1463 | |
GenBank mRNA | AB017027.1 | - | 3, 4 | 1464 | |
BC087730.1 | - | 3, 4 | 1388 | ||
BC138630.1 | - | 3, 4 | 1212 | 8861650 | |
BC138633.1 | - | 3, 4 | 1212 | 8861653 |