Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025768.2 | NP_080044.2 | 5, 6 | 671 | |
GenBank mRNA | AF329833.1 | - | 3, 4 | 459 | |
AK007905.1 | - | 5, 6 | 676 | ||
AK017197.1 | - | 5, 6 | 635 | ||
AK017251.1 | - | 6, 7 | 633 | ||
AK082863.1 | - | 5, 6 | 665 | ||
AK149322.1 | - | 668 | |||
BC029096.1 | - | 5, 6 | 677 | 3500563 |