Gene Transcripts

Gene Symbol
Vps11
Entrez Gene ID
71732
Gene Name
vacuolar protein sorting 11 (yeast)
Gene Aliases
1200011A11Rik
Chromosome Location
Chr. 9: 44347881 - 44361675 on Build GRCm38
UniGene ID
Mm.295013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027889.1 NP_082165.1 14 2347
GenBank mRNA AK004695.1 - 14 2322
AK032484.1 - 14 2312
AK037658.1 - 12 2802
AK090030.1 - 14 2328
AK153963.1 - 14 2320
AK162897.1 - 14 2322
AK179446.1 - 2 54
BC029004.1 - 14 2347 5360111

View Full Product Details