Gene Transcripts
- Gene Symbol
- Spag17
- Entrez Gene ID
- 74362
- Gene Name
- sperm associated antigen 17
- Gene Aliases
- 4931427F14Rik, PF6, Spag17-ps
- Chromosome Location
- Chr. 3: 99885417 - 100143322 on Build GRCm38
- UniGene ID
- Mm.55847
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028892.4 |
NP_083168.3
|
29, 30 |
4424 |
|
GenBank mRNA |
AY792594.1 |
- |
28, 29 |
3975 |
|
View Full Product Details