Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172601.3 | NP_766189.1 | 7 | 683 | |
GenBank mRNA | AB232586.1 | - | 7 | 529 | |
AK005230.1 | - | 7 | 586 | ||
AK038393.1 | - | 7 | 618 | ||
AK044202.1 | - | 7 | 560 | ||
AK160249.1 | - | 7 | 834 | ||
AK160351.1 | - | 7 | 549 | ||
BC033312.1 | - | 7 | 580 | 4481516 | |
BC046334.1 | - | 7 | 625 | 6492838 | |
BC054719.1 | - | 7 | 616 | ||
BC061513.1 | - | 3 | 632 |