Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178646.4 | NP_848761.2 | 1 | 476 | |
GenBank mRNA | AK036241.1 | - | 1 | 449 | |
AK051177.1 | - | 1 | 489 | ||
AK086663.1 | - | 1 | 452 | ||
AK165093.1 | - | 1 | 452 | ||
BC099838.1 | - | 1 | 476 | 5703010 | |
BC138289.1 | - | 1 | 450 | 8861308 |