Gene Transcripts

Gene Symbol
Skint4
Entrez Gene ID
320640
Gene Name
selection and upkeep of intraepithelial T cells 4
Gene Aliases
9530098N22Rik
Chromosome Location
Chr. 4: 112072016 - 112168076 on Build GRCm38
UniGene ID
Mm.253775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178786.4 NP_848901.2 2, 3 335
GenBank mRNA AK028550.1 - 2, 3 386
AK028795.1 - 2, 3 336
AK035748.1 - 2, 3 253
AK162813.1 - 3, 4 350
EF494897.1 - 2, 3 134
EF494898.1 - 2, 3 134

View Full Product Details