Gene Transcripts

Gene Symbol
Ric3
Entrez Gene ID
320360
Gene Name
resistance to inhibitors of cholinesterase 3 homolog (C. elegans)
Gene Aliases
E130307J04Rik
Chromosome Location
Chr. 7: 109034319 - 109083389 on Build GRCm38
UniGene ID
Mm.71007
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038624.1 NP_001033713.1 3, 4 462
NM_178780.3 NP_848895.2 3, 4 462
GenBank mRNA AK053760.1 - 3, 4 443
AK082275.1 - 3, 4 443
AK134663.1 - 3, 4 444
AK138641.1 - 3, 4 461
AK149334.1 - 3, 4 450
BC059258.1 - 2, 3 204 6408852

View Full Product Details