Gene Transcripts

Gene Symbol
Snrpd1
Entrez Gene ID
20641
Gene Name
small nuclear ribonucleoprotein D1
Gene Aliases
AA407109, AL023031, SMD1
Chromosome Location
Chr. 18: 10617796 - 10628230 on Build GRCm38
UniGene ID
Mm.603
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009226.4 NP_033252.1 2, 3 216
GenBank mRNA AK005517.1 - 2, 3 206
AK007674.1 - 2, 3 217
AK011455.1 - 2, 3 210
AK013075.1 - 2, 3 183
AK134582.1 - 2, 3 210
AK135979.1 - 2, 3 206
BC024875.1 - 2, 3 202 4009014
M58558.1 - 2, 3 145

View Full Product Details