Gene Transcripts

Gene Symbol
Krtap19-5
Entrez Gene ID
16704
Gene Name
keratin associated protein 19-5
Gene Aliases
Krtap8-2
Chromosome Location
Chr. 16: 88895968 - 88896449 on Build GRCm38
UniGene ID
Mm.3503
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010676.2 NP_034806.2 1 132
GenBank mRNA AA717139.1 - 1 140
D86422.1 - 1 143

View Full Product Details