Gene Transcripts
- Gene Symbol
- Pink1
- Entrez Gene ID
- 68943
- Gene Name
- PTEN induced putative kinase 1
- Gene Aliases
- 1190006F07Rik, AU042772, AW557854, BRPK, mFLJ00387
- Chromosome Location
- Chr. 4: 138313409 - 138326296 on Build GRCm38
- UniGene ID
- Mm.18539
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026880.2 |
NP_081156.2
|
1, 2 |
463 |
|
GenBank mRNA |
AB053476.1 |
- |
1, 2 |
395 |
|
AF316872.1 |
- |
1, 2 |
441 |
|
AK050322.1 |
- |
1, 2 |
443 |
|
AK155474.1 |
- |
1, 2 |
449 |
|
AK165480.1 |
- |
1, 2 |
442 |
|
AK215426.1 |
- |
1, 2 |
24 |
|
AK220318.1 |
- |
1, 2 |
417 |
|
BC018561.1 |
- |
1, 2 |
289 |
|
BC018594.1 |
- |
1, 2 |
426 |
|
BC067066.1 |
- |
1, 2 |
431 |
30542123 |
BC069968.1 |
- |
2, 3 |
321 |
|
View Full Product Details