Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190453.1 | NP_001177382.1 | 4 | 425 | |
NM_001190454.1 | NP_001177383.1 | 3 | 434 | ||
NM_027151.2 | NP_081427.1 | 3 | 419 | ||
GenBank mRNA | AK144464.1 | - | 3 | 295 | |
AK144505.1 | - | 3 | 295 | ||
AK164021.1 | - | 3 | 296 | ||
BC004613.1 | - | 3 | 282 | 3585310 |