Gene Transcripts

Gene Symbol
Golm1
Entrez Gene ID
105348
Gene Name
golgi membrane protein 1
Gene Aliases
2310001L02Rik, AW125446, D030064E01Rik, GP73, Golph2, PSEC0257
Chromosome Location
Chr. 13: 59634996 - 59675784 on Build GRCm38
UniGene ID
Mm.171335
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035122.2 NP_001030294.1 3, 4 387
NM_027307.4 NP_081583.3 4, 5 523
GenBank mRNA AK009072.1 - 2, 3 194
AK133865.1 - 3, 4 389
AK167646.1 - 3, 4 364
BC011152.1 - 3, 4 348 4188434
BC098486.1 - 4, 5 519 30024953
CD552063.1 - 4, 5 484
CK391733.1 - 3, 4 387

View Full Product Details