Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027654.2 | NP_081930.1 | 2 | 458 | |
GenBank mRNA | AB047007.1 | - | 2 | 508 | |
AK016709.1 | - | 2 | 480 | ||
AK132448.1 | - | 2 | 504 | ||
AK155067.1 | - | 2 | 520 | ||
BC016195.1 | - | 2 | 458 | 4488614 | |
BC089460.1 | - | 2 | 497 | 30632537 |