Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001113367.1 | NP_001106838.1 | 3, 4 | 469 | |
NM_029267.3 | NP_083543.2 | 3, 4 | 414 | ||
GenBank mRNA | AF272859.1 | - | 3, 4 | 277 | |
AK016125.1 | - | 3, 4 | 415 | ||
AK016403.1 | - | 3, 4 | 469 |