Gene Transcripts

Gene Symbol
Il17c
Entrez Gene ID
234836
Gene Name
interleukin 17C
Gene Aliases
IL-17C
Chromosome Location
Chr. 8: 122422085 - 122423639 on Build GRCm38
UniGene ID
Mm.222808
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145834.3 NP_665833.3 1 237
GenBank mRNA AF458061.1 - 1 237
BC145853.1 - 1 300 40131156

View Full Product Details