Gene Transcripts

Gene Symbol
Hist4h4
Entrez Gene ID
320332
Gene Name
histone cluster 4, H4
Gene Aliases
B130044J01Rik
Chromosome Location
Chr. 6: 136803993 - 136804431 on Build GRCm38
UniGene ID
Mm.246720
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175652.3 NP_783583.1 1 240
GenBank mRNA AK045187.1 - 235
BC111813.1 - 1 187

View Full Product Details