Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177041.3 | NP_796015.2 | 3 | 374 | |
GenBank mRNA | AK044501.1 | - | 383 | ||
AK161929.1 | - | 3 | 418 | ||
BC025817.1 | - | 3 | 327 | 5099127 | |
BC046769.1 | - | 3 | 374 | 6809196 |