Gene Transcripts

Gene Symbol
Cyp4a12a
Entrez Gene ID
277753
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 12a
Gene Aliases
BC025936, Cyp4a12
Chromosome Location
Chr. 4: 115299046 - 115332815 on Build GRCm38
UniGene ID
Mm.276106
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC014721.1 - 9, 10 1252
BC025936.1 - 9, 10 1246 5097698
BC026582.1 - 9, 10 1252 4971335
BC031141.1 - 9, 10 1227 4972684
BC033924.1 - 9, 10 1236 5101821

View Full Product Details