Gene Transcripts
- Gene Symbol
- Fam26e
- Entrez Gene ID
- 103511
- Gene Name
- family with sequence similarity 26, member E
- Gene Aliases
- BB146404
- Chromosome Location
- Chr. 10: 34091352 - 34096519 on Build GRCm38
- UniGene ID
- Mm.66334
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK085925.1 |
- |
2 |
973 |
|
View Full Product Details