Gene Transcripts

Gene Symbol
Klhl2
Entrez Gene ID
77113
Gene Name
kelch-like 2, Mayven
Gene Aliases
6030411N21Rik, ABP-KELCH, AU020744, Mav
Chromosome Location
Chr. 8: 64739675 - 64793152 on Build GRCm38
UniGene ID
Mm.350347
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178633.3 NP_848748.2 3170
GenBank mRNA AK032116.1 - 2718
AK079387.1 - 2916
AK167903.1 - 2961

View Full Product Details