Gene Transcripts

Gene Symbol
Foxl1
Entrez Gene ID
14241
Gene Name
forkhead box L1
Gene Aliases
FREAC7, Fkh6, fkh-6
Chromosome Location
Chr. 8: 121127685 - 121130644 on Build GRCm38
UniGene ID
Mm.5111
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008024.2 NP_032050.2 1 443
GenBank mRNA AB221616.1 - 1 166
BC137805.1 - 1 188 8860824
BC137806.1 - 1 188 8860825

View Full Product Details